Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080850.2 | NP_001074319.1 | 15 | 2205 | ||
XM_011542067.2 | XP_011540369.1 | 16 | 3137 | |||
XM_011542068.2 | XP_011540370.1 | 16 | 3142 | |||
XM_011542069.2 | XP_011540371.1 | 15 | 2996 | |||
XM_011542070.2 | XP_011540372.1 | 14 | 2163 | |||
XM_011542071.2 | XP_011540373.1 | 13 | 2572 | |||
XM_017002225.1 | XP_016857714.1 | 15 | 2222 | |||
XM_017002226.1 | XP_016857715.1 | 16 | 2533 | |||
GenBank mRNA | AL832217.1 | - | 22 | 3238 | ||
AY639646.1 | - | 14 | 2030 |