Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTDSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290225.1 | NP_001277154.1 | 5 | 859 | ||
NM_014754.2 | NP_055569.1 | 7 | 1029 | |||
GenBank mRNA | AK293036.1 | - | 7 | 1013 | ||
AK293513.1 | - | 3 | 339 | |||
BC002376.2 | - | 5 | 832 | 2961075 | ||
BC004192.2 | - | 7 | 979 | 3509418 | ||
BC004390.1 | - | 7 | 979 | 3634879 | ||
BC004502.2 | - | 3 | 450 | |||
D14694.1 | - | 7 | 976 |