Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM63A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014698.2 | NP_055513.2 | 1 | 349 | ||
XM_011544330.2 | XP_011542632.1 | 1 | 349 | |||
XM_011544331.2 | XP_011542633.1 | 1 | 32 | |||
XM_011544332.2 | XP_011542634.1 | 1 | 32 | |||
GenBank mRNA | DA127076.1 | - | 1 | 350 |