Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ALG13 SIRNA Assays ›
Gene Symbol:
ALG13
Gene Name:
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases:
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
Chromosome Location:
Chr.X: 111680741 - 111764164 on Build GRCh38
UniGene ID:
Hs.443061
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s59079
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ALG13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099922.2
NP_001093392.1
8
1080
NM_001257230.1
NP_001244159.1
8
976
NM_001257231.1
NP_001244160.1
8
1094
NM_001257234.1
NP_001244163.1
8
1049
NM_001257237.1
NP_001244166.1
8
1153
NM_001324292.1
NP_001311221.1
8
1080
NM_001324293.1
NP_001311222.1
8
1153
XM_006724693.2
XP_006724756.1
8
1453
XM_006724697.3
XP_006724760.2
8
1353
XM_006724698.3
XP_006724761.2
8
1478
XM_011531028.2
XP_011529330.1
8
1454
XM_011531030.1
XP_011529332.1
8
1452
XM_011531031.1
XP_011529333.1
8
1153
XM_011531032.1
XP_011529334.1
8
959
XM_011531033.2
XP_011529335.1
8
999
XM_011531034.2
XP_011529336.1
6
693
XM_017029846.1
XP_016885335.1
8
1455
XM_017029847.1
XP_016885336.1
8
1454
XM_017029848.1
XP_016885337.1
8
1452
XM_017029849.1
XP_016885338.1
8
1453
XM_017029850.1
XP_016885339.1
8
1452
XM_017029851.1
XP_016885340.1
9
1361
XM_017029853.1
XP_016885342.1
5
377
XM_017029855.1
XP_016885344.1
2
827
GenBank mRNA
AK300394.1
-
8
918
AK302729.1
-
8
1021
AK302890.1
-
8
997
AK304712.1
-
8
970
AK316306.1
-
8
1022
AK316522.1
-
8
1097
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
transferase
Gene Ontology Categories:
Function(s)
Process(es)
dolichol-linked oligosaccharide biosynthetic process
proteolysis
N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase activity
thiol-dependent ubiquitinyl hydrolase activity
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0