Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052847.2 | NP_443079.1 | 4, 5 | 317 | ||
XM_017026606.1 | XP_016882095.1 | 4, 5 | 426 | |||
GenBank mRNA | AB010414.1 | - | 2, 3 | 158 | ||
AF493874.1 | - | 1, 2 | 80 | |||
AK311750.1 | - | 4, 5 | 254 | |||
BC014466.1 | - | 4, 5 | 278 | 4861393 | ||
BC053630.1 | - | 4, 5 | 234 | 6469872 | ||
DA106844.1 | - | 4, 5 | 317 | |||
DA247608.1 | - | 4, 5 | 261 |