Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015963.5 | NP_057047.4 | 2 | 998 | ||
XM_005247016.4 | XP_005247073.3 | 2 | 1231 | |||
XM_011511291.2 | XP_011509593.1 | 2 | 800 | |||
XM_017004256.1 | XP_016859745.1 | 2 | 792 | |||
GenBank mRNA | AF258556.1 | - | 2 | 750 | ||
AK001216.1 | - | 1 | 45 | |||
BC000247.1 | - | 1 | 440 | |||
BC009439.1 | - | 1 | 536 | |||
BC069235.1 | - | 2 | 928 | 6645198 |