Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100595.1 | NP_001094065.1 | 2 | 552 | ||
NM_182620.3 | NP_872426.1 | 2 | 377 | |||
XM_011524745.2 | XP_011523047.1 | 2 | 191 | |||
GenBank mRNA | AK022820.1 | - | 2 | 109 | ||
AK056473.1 | - | 2 | 129 | |||
BC017873.1 | - | 2 | 138 | 4685972 | ||
BC106003.1 | - | 2 | 102 | 6143473 | ||
DA133216.1 | - | 2 | 290 |