Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000825.3 | NP_000816.4 | 1 | 1822 | ||
NM_001083111.1 | NP_001076580.1 | 2 | 953 | |||
GenBank mRNA | BC126437.1 | - | 1 | 644 | 8992153 | |
BC126463.1 | - | 1 | 643 | 8992179 | ||
M12578.1 | - | 2 | 144 | |||
X01059.1 | - | 1 | 1186 |