Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other VOPP1 SIRNA Assays ›
Gene Symbol:
VOPP1
Gene Name:
vesicular, overexpressed in cancer, prosurvival protein 1
Gene Aliases:
ECOP, GASP
Chromosome Location:
Chr.7: 55434406 - 55572520 on Build GRCh38
UniGene ID:
Hs.488307
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s59350
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: VOPP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284282.1
NP_001271211.1
2
165
NM_001284283.1
NP_001271212.1
2
99
NM_001284284.1
NP_001271213.1
2
260
NM_001321242.1
NP_001308171.1
2
180
NM_001321243.1
NP_001308172.1
2
294
NM_001321244.1
NP_001308173.1
3
366
NM_001321246.1
NP_001308175.1
2
294
NM_001321247.1
NP_001308176.1
2
294
NM_001321248.1
NP_001308177.1
2
121
NM_001321249.1
NP_001308178.1
2
182
NM_001321250.1
NP_001308179.1
2
204
NM_001321251.1
NP_001308180.1
2
190
NM_030796.4
NP_110423.3
2
294
XM_011515539.1
XP_011513841.1
2
236
XM_011515540.2
XP_011513842.1
2
442
XM_011515541.2
XP_011513843.1
2
1135
XM_011515546.2
XP_011513848.1
2
964
GenBank mRNA
AB097004.1
-
2
153
AF395824.1
-
1
59
AK075391.1
-
2
228
AK090825.1
-
2
204
AK092992.1
-
2
238
AK225755.1
-
2
239
AK309288.1
-
2
165
AK314338.1
-
2
187
AK315882.1
-
2
92
AL136610.1
-
1
107
BC016650.1
-
1
186
4342536
BI597655.1
-
220
CN299440.1
-
2
300
CR533483.1
-
2
98
DA257111.1
-
2
187
DA530391.1
-
2
121
DA732543.1
-
3
133
DA772419.1
-
2
190
DA814621.1
-
2
99
DA991268.1
-
2
181
DB496308.1
-
2
299
HY032103.1
-
2
183
HY057980.1
-
2
215
HY073578.1
-
2
264
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
signal transduction
signal transducer activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0