Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100505585 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135626.1 | 2 | 610 |
Gene Symbol: ARHGEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832294.1 | - | 21 | 2583 |