Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123041.2 | NP_001116513.2 | 3 | 1546 | ||
XM_011534069.1 | XP_011532371.1 | 3 | 2467 | |||
GenBank mRNA | BC074751.2 | - | 2 | 1100 | 30915210 | |
BC126452.1 | - | 2 | 1095 | 8992168 | ||
BC136396.1 | - | 2 | 1095 | 9020383 | ||
U03882.1 | - | 2 | 1100 | |||
X95583.1 | - | 1 | 1617 |
Set Membership: |
G-protein coupled receptor |