Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002079.2 | NP_002070.1 | 3 | 577 | ||
GenBank mRNA | AF052153.1 | - | 3 | 397 | ||
AK293483.1 | - | 3 | 299 | |||
AK301916.1 | - | 3 | 475 | |||
AK312684.1 | - | 3 | 419 | |||
AY513279.1 | - | 3 | 371 | |||
BC000498.2 | - | 3 | 397 | 2820849 | ||
M37400.1 | - | 3 | 393 |