Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286220.1 | NP_001273149.1 | 7 | 921 | ||
NM_002080.3 | NP_002071.2 | 8 | 1050 | |||
GenBank mRNA | AK098313.1 | - | 8 | 972 | ||
AK223271.1 | - | 8 | 985 | |||
AK290847.1 | - | 8 | 983 | |||
AK295993.1 | - | 7 | 890 | |||
AK302249.1 | - | 7 | 891 | |||
AK307598.1 | - | 8 | 937 | |||
AL516773.3 | - | 7 | 884 | |||
BC000525.2 | - | 8 | 973 | 2961181 | ||
M22632.1 | - | 8 | 929 |