Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000175.3 | NP_000166.2 | 12 | 1115 | ||
NM_001184722.1 | NP_001171651.1 | 12 | 1178 | |||
NM_001289789.1 | NP_001276718.1 | 13 | 1262 | |||
NM_001289790.1 | NP_001276719.1 | 11 | 1031 | |||
XM_005258764.2 | XP_005258821.1 | 13 | 1081 | |||
XM_006723148.2 | XP_006723211.1 | 13 | 1032 | |||
XM_011526754.2 | XP_011525056.1 | 14 | 1497 | |||
GenBank mRNA | AK025421.1 | - | 1 | 713 | ||
AK129884.1 | - | 13 | 1046 | |||
AK130102.1 | - | 11 | 810 | |||
AK293446.1 | - | 11 | 907 | |||
AK294396.1 | - | 12 | 1126 | |||
BC004982.1 | - | 12 | 983 | 2906270 | ||
K03515.1 | - | 12 | 990 |