Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286099.1 | NP_001273028.1 | 3 | 843 | ||
NM_005284.4 | NP_005275.1 | 2 | 734 | |||
GenBank mRNA | AK294648.1 | - | 2 | 684 | ||
AK295250.1 | - | 3 | 788 | |||
BC043414.2 | - | 1 | 522 | 5294842 | ||
BC117298.1 | - | 1 | 620 | 40125849 | ||
BC126301.1 | - | 1 | 622 | 8992017 | ||
CF147782.1 | - | 1 | 572 | |||
HQ709190.1 | - | 1 | 504 | |||
HQ709191.1 | - | 1 | 504 |
Set Membership: |
G-protein coupled receptor |