Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078170.2 | NP_001071638.2 | 20 | 4756 | ||
XM_017004845.1 | XP_016860334.1 | 24 | 5207 | |||
XM_017004846.1 | XP_016860335.1 | 20 | 4878 | |||
XM_017004847.1 | XP_016860336.1 | 20 | 4875 | |||
XM_017004848.1 | XP_016860337.1 | 24 | 5207 | |||
XM_017004849.1 | XP_016860338.1 | 24 | 5207 | |||
XM_017004850.1 | XP_016860339.1 | 24 | 5207 | |||
GenBank mRNA | AK298885.1 | - | 1 | 206 | ||
AK300276.1 | - | 1 | 1543 | |||
AK300352.1 | - | 9 | 3096 | |||
AK302429.1 | - | 20 | 4756 | |||
AY354203.1 | - | 7 | 2732 |
Gene Symbol: RGPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024457.3 | NP_001019628.3 | 20 | 4710 | ||
XM_011532845.2 | XP_011531147.1 | 21 | 4909 | |||
XM_011532849.2 | XP_011531151.1 | 21 | 4909 | |||
XM_011532850.1 | XP_011531152.1 | 21 | 4909 | |||
XM_011532851.2 | XP_011531153.1 | 21 | 4909 | |||
XM_011532852.2 | XP_011531154.1 | 21 | 4909 | |||
XM_017004103.1 | XP_016859592.1 | 21 | 4909 | |||
XM_017004104.1 | XP_016859593.1 | 20 | 5026 | |||
XM_017004105.1 | XP_016859594.1 | 20 | 4874 | |||
XM_017004106.1 | XP_016859595.1 | 20 | 4993 | |||
XM_017004107.1 | XP_016859596.1 | 21 | 4909 | |||
GenBank mRNA | AK303443.1 | - | 6 | 2626 | ||
CR749330.1 | - | 20 | 4710 | |||
DC378175.1 | - | 1 | 228 |