Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005876.4 | NP_005867.3 | 25 | 5302 | ||
XM_005246237.2 | XP_005246294.1 | 27 | 5030 | |||
XM_005246239.2 | XP_005246296.1 | 27 | 4845 | |||
XM_005246240.2 | XP_005246297.1 | 20 | 3047 | |||
XM_005246241.1 | XP_005246298.1 | 20 | 2941 | |||
XM_005246242.4 | XP_005246299.1 | 20 | 3066 | |||
XM_006712189.3 | XP_006712252.1 | 26 | 5035 | |||
XM_006712193.3 | XP_006712256.1 | 20 | 6166 | |||
XM_011510479.2 | XP_011508781.1 | 26 | 5468 | |||
XM_011510483.2 | XP_011508785.2 | 26 | 5065 | |||
XM_017003157.1 | XP_016858646.1 | 26 | 5065 | |||
XM_017003158.1 | XP_016858647.1 | 22 | 4641 | |||
XM_017003159.1 | XP_016858648.1 | 23 | 4824 | |||
XM_017003160.1 | XP_016858649.1 | 10 | 1237 | |||
GenBank mRNA | AB037718.1 | - | 16 | 2359 | ||
AY603755.1 | - | 24 | 4915 |
Set Membership: |
G-protein coupled receptor |