Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040441.2 | NP_001035531.2 | 4, 5 | 1511 | ||
GenBank mRNA | AF548353.1 | - | 3, 4 | 1043 | ||
BC015067.1 | - | 2, 3 | 385 | |||
BC015239.1 | - | 4, 5 | 1179 | 3914043 | ||
BC029654.1 | - | 2, 3 | 431 |