Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000314.6 | NP_000305.3 | 9 | 2184 | ||
NM_001304717.2 | NP_001291646.2 | 9 | 2184 | |||
NM_001304718.1 | NP_001291647.1 | 9 | 2298 | |||
GenBank mRNA | AK313581.1 | - | 9 | 1345 | ||
BC005821.2 | - | 9 | 1989 | 3937787 | ||
CR450306.1 | - | 9 | 1153 | |||
JF268690.1 | - | 9 | 1316 | |||
KJ534922.1 | - | 9 | 1153 | |||
U92436.1 | - | 9 | 2187 | |||
U93051.1 | - | 9 | 1153 | |||
U96180.1 | - | 9 | 1957 |
Gene Symbol: PTENP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023917.1 | 1 | 1922 | |||
GenBank mRNA | AF019083.1 | - | 1 | 1153 | ||
BC038293.2 | - | 1 | 1406 |