Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017551.2 | NP_060021.1 | 10 | 1543 | ||
XM_011539720.2 | XP_011538022.1 | 10 | 1540 | |||
GenBank mRNA | AB033046.1 | - | 7 | 790 | ||
AK127168.1 | - | 10 | 1528 | |||
AK293104.1 | - | 10 | 1501 | |||
AK302192.1 | - | 3 | 315 | |||
BC039263.1 | - | 10 | 1543 | 5273869 |