Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005264292.2 | XP_005264349.1 | 2 | 420 | ||
XM_006712007.3 | XP_006712070.1 | 2 | 162 | |||
XM_011532823.2 | XP_011531125.1 | 2 | 379 |