Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WHAMMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003521.1 | 5 | 1131 | |||
GenBank mRNA | AL833132.1 | - | 4 | 583 | ||
BC018097.1 | - | 4 | 450 | |||
BC044610.1 | - | 4 | 583 | |||
BC048987.1 | - | 5 | 1191 | |||
BC093703.1 | - | 2 | 188 | |||
BC093705.1 | - | 2 | 188 | |||
DQ309039.1 | - | 4 | 440 | |||
DQ309040.1 | - | 5 | 1131 |
Gene Symbol: WHAMM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080435.2 | NP_001073904.1 | 4 | 1101 | ||
XM_005272421.4 | XP_005272478.1 | 4 | 717 | |||
XM_005272422.3 | XP_005272479.1 | 4 | 464 | |||
XM_005272423.4 | XP_005272480.1 | 4 | 703 | |||
XM_011521232.2 | XP_011519534.1 | 2 | 913 | |||
XM_017021923.1 | XP_016877412.1 | 4 | 2550 | |||
GenBank mRNA | AB075851.1 | - | 4 | 1087 | ||
AK126887.1 | - | 4 | 1102 |
Gene Symbol: WHAMMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036650.1 | 3 | 754 | |||
GenBank mRNA | BC117556.1 | - | 754 | |||
BI910970.1 | - | 2 | 190 |
Gene Symbol: WHAMMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026589.1 | 5 | 1155 | |||
GenBank mRNA | BC035099.1 | - | 5 | 705 | ||
BC066982.1 | - | 5 | 879 | |||
BC150524.1 | - | 3 | 384 |