Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319238.1 | NP_001306167.1 | 1 | 238 | ||
NM_001319239.1 | NP_001306168.1 | 1 | 238 | |||
XM_005245643.2 | XP_005245700.1 | 1 | 238 | |||
XM_005245644.3 | XP_005245701.1 | 1 | 126 | |||
XM_006711673.2 | XP_006711736.1 | 1 | 138 | |||
XM_011510199.1 | XP_011508501.1 | 1 | 166 | |||
XM_011510200.1 | XP_011508502.1 | 1 | 126 | |||
XM_011510202.1 | XP_011508504.1 | 1 | 105 | |||
XM_017002956.1 | XP_016858445.1 | 1 | 111 | |||
XM_017002957.1 | XP_016858446.1 | 1 | 121 | |||
XM_017002959.1 | XP_016858448.1 | 1 | 121 | |||
XM_017002960.1 | XP_016858449.1 | 1 | 111 | |||
XM_017002961.1 | XP_016858450.1 | 1 | 166 | |||
XM_017002962.1 | XP_016858451.1 | 1 | 126 | |||
XM_017002964.1 | XP_016858453.1 | 1 | 166 | |||
GenBank mRNA | AK095858.1 | - | 1 | 105 | ||
AK296983.1 | - | 1 | 95 | |||
BC038513.2 | - | 1 | 69 | |||
CR627439.1 | - | 1 | 116 | |||
DB066230.1 | - | 1 | 238 |