Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267585.1 | NP_001254514.1 | 12 | 2375 | ||
NM_001267586.1 | NP_001254515.1 | 11 | 2216 | |||
NR_051988.1 | 12 | 3021 | ||||
XM_011523625.1 | XP_011521927.1 | 11 | 2528 | |||
XM_017024020.1 | XP_016879509.1 | 11 | 2528 | |||
XM_017024021.1 | XP_016879510.1 | 11 | 2491 | |||
GenBank mRNA | AK310383.1 | - | 12 | 2960 | ||
AL136822.1 | - | 11 | 2154 | |||
AY729024.1 | - | 12 | 2143 | |||
BC028364.2 | - | 11 | 2449 | 4827344 | ||
BI828401.1 | - | 2 | 257 |