Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002156.4 | NP_002147.2 | 11, 12 | 1706 | ||
NM_199440.1 | NP_955472.1 | 11, 12 | 1686 | |||
GenBank mRNA | AK297305.1 | - | 1633 | |||
AK298632.1 | - | 11, 12 | 1629 | |||
AK298844.1 | - | 10, 11 | 1457 | |||
AK312240.1 | - | 11, 12 | 1639 | |||
BC002676.2 | - | 11, 12 | 1616 | 3609039 | ||
BC003030.1 | - | 11, 12 | 1615 | 2821157 | ||
BC047350.1 | - | 10, 11 | 1776 | |||
BC067082.1 | - | 11, 12 | 1614 | 4025725 | ||
BC073746.1 | - | 11, 12 | 1614 | 6372068 | ||
BX647145.1 | - | 10, 11 | 2371 | |||
EU719626.1 | - | 10, 11 | 1569 | |||
KU178139.1 | - | 10, 11 | 1569 | |||
KU178140.1 | - | 5, 6 | 820 | |||
M22382.1 | - | 11, 12 | 1614 | |||
M34664.1 | - | 11, 12 | 1593 |