Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROCK1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033770.1 | 4 | 813 | |||
GenBank mRNA | BC041849.1 | - | 4 | 813 | ||
DB095042.1 | - | 4 | 450 |
Gene Symbol: ROCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005406.2 | NP_005397.1 | 32 | 4877 | ||
XM_011526137.2 | XP_011524439.1 | 21 | 3111 | |||
GenBank mRNA | AU121527.1 | - | 1 | 80 | ||
BC113114.1 | - | 32 | 3952 | 7961989 | ||
U43195.1 | - | 32 | 3936 |