Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043229.1 | NP_001036694.1 | 3 | 549 | ||
XM_005274232.4 | XP_005274289.1 | 2 | 918 | |||
GenBank mRNA | AK297138.1 | - | 3 | 549 | ||
BC147001.1 | - | 2 | 428 | 9056929 | ||
BC147011.1 | - | 2 | 428 | 9056939 | ||
BC171895.1 | - | 2 | 470 | |||
BX348415.2 | - | 1 | 116 | |||
BY796224.2 | - | 2 | 505 |