Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077620.2 | NP_001071088.1 | 5 | 787 | ||
NR_033357.1 | 5 | 858 | ||||
XM_017025013.1 | XP_016880502.1 | 5 | 2805 | |||
XM_017025014.1 | XP_016880503.1 | 5 | 2805 | |||
GenBank mRNA | AK054729.1 | - | 1 | 327 | ||
AK125617.1 | - | 5 | 858 | |||
BC043413.1 | - | 5 | 655 |