Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TVP23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079512.3 | NP_001072980.1 | 4, 5 | 633 | ||
NM_001318873.1 | NP_001305802.1 | 4, 5 | 1084 | |||
NR_134901.1 | 5, 6 | 703 | ||||
NR_134902.1 | 4, 5 | 633 | ||||
NR_134903.1 | 4, 5 | 595 | ||||
NR_134904.1 | 4, 5 | 558 | ||||
NR_134905.1 | 3, 4 | 488 | ||||
NR_134906.1 | 3, 4 | 488 | ||||
XM_006720944.3 | XP_006721007.1 | 4, 5 | 741 | |||
XM_011522654.2 | XP_011520956.1 | 4, 5 | 741 | |||
XM_017023649.1 | XP_016879138.1 | 4, 5 | 741 | |||
XM_017023650.1 | XP_016879139.1 | 4, 5 | 941 | |||
XM_017023651.1 | XP_016879140.1 | 4, 5 | 369 | |||
GenBank mRNA | AK124106.1 | - | 4, 5 | 941 | ||
AK294418.1 | - | 4, 5 | 624 | |||
AL833564.1 | - | 2, 3 | 143 | |||
BC050569.1 | - | 4, 5 | 408 | |||
BC146882.1 | - | 4, 5 | 439 | 9056810 | ||
BC146886.1 | - | 4, 5 | 439 | 9056814 | ||
BC171807.1 | - | 3, 4 | 294 | |||
BC171816.1 | - | 3, 4 | 294 | |||
BC171817.1 | - | 4, 5 | 364 | 9054471 | ||
BC171819.1 | - | 5, 6 | 509 | |||
CN301583.1 | - | 4, 5 | 603 |