Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289791.1 | NP_001276720.1 | 4 | 1664 | ||
NM_009597.1 | NP_033727.1 | 6 | 1142 | |||
GenBank mRNA | AB208022.1 | - | 4 | 1048 | ||
BC067025.1 | - | 6 | 1142 | 6850826 |