Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007385.3 | NP_031411.1 | 3 | 334 | ||
GenBank mRNA | BB872041.1 | - | 3 | 332 | ||
BC024657.1 | - | 3 | 290 | 4196217 | ||
BC100593.1 | - | 3 | 333 |