Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102404.1 | NP_001095874.1 | 4 | 554 | ||
NM_001102405.1 | NP_001095875.1 | 4 | 559 | |||
NM_007388.3 | NP_031414.1 | 4 | 501 | |||
GenBank mRNA | AK008391.1 | - | 3 | 281 | ||
AK010865.1 | - | 4 | 525 | |||
AK011915.1 | - | 4 | 525 | |||
BC012911.1 | - | 4 | 453 | 3588062 | ||
BC019160.1 | - | 4 | 517 | 3596147 | ||
BC029644.1 | - | 4 | 453 | 3584530 | ||
S70805.1 | - | 3 | 177 |