Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009602.4 | NP_033732.2 | 6 | 1575 | ||
GenBank mRNA | AF145286.1 | - | 6 | 1514 | ||
AF299083.1 | - | 6 | 1369 | |||
AK049722.1 | - | 6 | 1548 | |||
AK051742.1 | - | 6 | 1557 | |||
AY574267.1 | - | 6 | 1349 | |||
BC065103.1 | - | 6 | 1528 | 6838257 |