Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272058.1 | NP_001258987.1 | 19 | 2486 | ||
NM_007401.3 | NP_031427.2 | 19 | 2486 | |||
GenBank mRNA | AK016574.1 | - | 19 | 2142 | ||
AK029519.1 | - | 19 | 2176 | |||
AK161262.1 | - | 19 | 2486 | |||
AK161417.1 | - | 19 | 2156 | |||
U22059.1 | - | 19 | 2150 |