Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291903.1 | NP_001278832.1 | 3 | 2404 | ||
NM_009623.2 | NP_033753.2 | 3 | 2404 | |||
GenBank mRNA | AK138244.1 | - | 3 | 2405 | ||
AK163443.1 | - | 3 | 2380 | |||
BC059010.1 | - | 3 | 1502 | |||
BC094231.1 | - | 3 | 1842 | |||
U85021.1 | - | 3 | 2264 |