Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291910.1 | NP_001278839.1 | 4 | 2258 | ||
NM_009624.3 | NP_033754.2 | 3 | 2481 | |||
GenBank mRNA | AK156940.1 | - | 4 | 2260 | ||
U30602.1 | - | 3 | 1788 | |||
Z50190.1 | - | 2 | 1757 |