Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007409.2 | NP_031435.1 | 8 | 1106 | ||
GenBank mRNA | AK146094.1 | - | 8 | 1107 | ||
BC013477.1 | - | 8 | 1106 | 4238555 | ||
BC054467.1 | - | 8 | 1086 | 6530906 | ||
M22611.1 | - | 3 | 365 |