Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007422.3 | NP_031448.2 | 12 | 1404 | ||
GenBank mRNA | AK004877.1 | - | 12 | 1357 | ||
AK010263.1 | - | 12 | 1379 | |||
AK028060.1 | - | 12 | 1404 | |||
AK148420.1 | - | 12 | 1386 | |||
AK170279.1 | - | 12 | 1390 | |||
AK216828.1 | - | 2 | 164 | |||
BC034755.1 | - | 1 | 918 | |||
BC040829.1 | - | 1 | 907 | |||
BC139404.1 | - | 12 | 1316 | 8862426 | ||
BC139417.1 | - | 12 | 1316 | 8862439 | ||
L24554.1 | - | 12 | 1371 |