Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRM8 SIRNA Assays ›
Gene Symbol:
GRM8
Gene Name:
glutamate metabotropic receptor 8
Gene Aliases:
GLUR8, GPRC1H, MGLUR8, mGlu8
Chromosome Location:
Chr.7: 126438598 - 127253294 on Build GRCh38
UniGene ID:
Hs.449625
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s6212
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRM8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000845.2
NP_000836.2
6
1522
NM_001127323.1
NP_001120795.1
7
1768
NR_028041.1
6
1522
XM_006715938.3
XP_006716001.1
7
1842
XM_011516091.1
XP_011514393.1
7
2020
XM_011516092.2
XP_011514394.1
7
2020
XM_011516094.1
XP_011514396.1
7
766
XM_011516095.2
XP_011514397.1
7
899
XM_011516101.2
XP_011514403.1
6
771
XM_011516102.2
XP_011514404.1
7
796
XM_011516103.2
XP_011514405.1
7
928
XM_011516108.2
XP_011514410.1
2
870
XM_017012074.1
XP_016867563.1
7
1621
XM_017012075.1
XP_016867564.1
7
972
XM_017012076.1
XP_016867565.1
7
990
XM_017012077.1
XP_016867566.1
6
844
XM_017012078.1
XP_016867567.1
7
869
XM_017012079.1
XP_016867568.1
6
792
XM_017012080.1
XP_016867569.1
5
1448
GenBank mRNA
AJ236921.1
-
6
1226
AJ236922.1
-
6
1226
AK123053.1
-
7
928
AK290197.1
-
7
1819
AK315203.1
-
7
1768
AY608335.1
-
6
1211
BC093725.1
-
6
1237
7939570
BC101675.1
-
6
1237
8069181
BC143344.1
-
6
1237
EU432125.1
-
6
1211
U92459.1
-
6
1268
U95025.1
-
6
1211
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
adenylate cyclase-inhibiting G-protein coupled glutamate receptor signaling pathway
visual perception
sensory perception of smell
negative regulation of cAMP biosynthetic process
regulation of synaptic transmission, glutamatergic
group III metabotropic glutamate receptor activity
G-protein coupled receptor activity
glutamate receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0