Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agxt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276710.1 | NP_001263639.1 | 7, 8 | 895 | ||
NM_016702.3 | NP_057911.2 | 7, 8 | 895 | |||
GenBank mRNA | AF027730.2 | - | 7, 8 | 890 | ||
AK149432.1 | - | 7, 8 | 818 | |||
BC025799.1 | - | 7, 8 | 859 | 5099627 |