Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172393.2 | NP_765981.2 | 2 | 1865 | ||
GenBank mRNA | AK144358.1 | - | 2 | 1388 |