Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035532.2 | NP_001030609.1 | 2 | 1620 | ||
NM_001035533.2 | NP_001030610.1 | 1 | 1561 | |||
NM_009649.3 | NP_033779.2 | 2 | 1620 | |||
GenBank mRNA | AF033274.1 | - | 1 | 1963 | ||
AF033275.1 | - | 1 | 1963 | |||
AF033276.1 | - | 1 | 1963 | |||
AK041873.1 | - | 2 | 1610 | |||
AK147420.1 | - | 2 | 1588 | |||
AK173063.1 | - | 1 | 2081 | |||
AK209521.1 | - | 1 | 67 | |||
BC003735.1 | - | 2 | 1615 | 3493391 |
Gene Symbol: Pakap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077020.1 | - | 3 | 2025 |