Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042542.2 | NP_001036007.1 | 5 | 2053 | ||
NM_009651.4 | NP_033781.2 | 5 | 2049 | |||
GenBank mRNA | BC130271.1 | - | 5 | 1981 | 40143193 | |
U07423.1 | - | 5 | 2022 | |||
U10341.1 | - | 5 | 2054 |