Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165894.1 | NP_001159366.1 | 10 | 1235 | ||
NM_009652.3 | NP_033782.1 | 10 | 1235 | |||
GenBank mRNA | AK040758.1 | - | 10 | 1041 | ||
AK154936.1 | - | 10 | 1016 | |||
BC066018.1 | - | 10 | 1235 | 5716837 | ||
BC115583.1 | - | 1 | 35 | |||
M94335.1 | - | 9 | 887 | |||
X65687.1 | - | 10 | 1148 |