Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009662.2 | NP_033792.1 | 6 | 972 | ||
GenBank mRNA | AK089344.1 | - | 6 | 926 | ||
AK136868.1 | - | 6 | 941 | |||
AK137481.1 | - | 6 | 930 | |||
AK169938.1 | - | 6 | 929 | |||
AK170016.1 | - | 6 | 829 | |||
AK170434.1 | - | 6 | 929 | |||
AK170879.1 | - | 6 | 928 | |||
AK171191.1 | - | 6 | 921 | |||
AK171413.1 | - | 6 | 929 | |||
BC139102.1 | - | 6 | 899 | 8862124 | ||
BC141213.1 | - | 6 | 899 | 9055825 | ||
L42198.1 | - | 6 | 818 |