Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox5ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004002.1 | - | 5 | 508 | ||
AK014153.1 | - | 2 | 149 | |||
AK075967.1 | - | 5 | 491 | |||
BC026209.1 | - | 5 | 495 | 1349819 | ||
M96554.1 | - | 5 | 440 |