Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007441.3 | NP_031467.1 | 2 | 671 | ||
GenBank mRNA | AK142019.1 | - | 2 | 597 | ||
U96109.1 | - | 2 | 674 |