Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009665.5 | NP_033795.1 | 8 | 1899 | ||
GenBank mRNA | AK146840.1 | - | 8 | 1898 | ||
AK167423.1 | - | 3 | 949 | |||
BC011110.1 | - | 8 | 1884 | 3672700 | ||
BC071220.1 | - | 8 | 1882 | 5012910 | ||
BC080791.1 | - | 8 | 1663 | 4014056 | ||
BC092072.1 | - | 8 | 1880 | 4505452 | ||
Z14986.1 | - | 8 | 1825 |
Gene Symbol: Amd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007444.3 | NP_031470.3 | 1 | 1901 |