Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110505.1 | NP_001103975.1 | 11 | 1572 | ||
NM_007446.2 | NP_031472.2 | 11 | 1681 | |||
GenBank mRNA | AA616210.1 | - | 4 | 466 | ||
AK029642.1 | - | 11 | 1604 | |||
BB462169.1 | - | 2 | 221 | |||
BC004698.1 | - | 7 | 915 | |||
BC009121.1 | - | 11 | 1555 | 3710941 | ||
J00356.1 | - | 10 | 1525 | |||
V00717.1 | - | 11 | 1570 | |||
V00719.1 | - | 11 | 1681 | |||
V00720.1 | - | 11 | 1572 |