Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110505.1 | NP_001103975.1 | 8 | 1155 | ||
NM_007446.2 | NP_031472.2 | 8 | 1264 | |||
GenBank mRNA | AA616210.1 | - | 1 | 47 | ||
AK029642.1 | - | 8 | 1187 | |||
BC004698.1 | - | 4 | 498 | |||
BC009121.1 | - | 8 | 1138 | 3710941 | ||
J00356.1 | - | 7 | 1108 | |||
V00717.1 | - | 8 | 1153 | |||
V00719.1 | - | 8 | 1264 | |||
V00720.1 | - | 8 | 1155 |